Thermo Scientific Chemicals

4-Carboxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53285.06
5 g, Each
Thermo Scientific Chemicals

4-Carboxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53285.06
5 g, Each
Quantity
Catalog number: H53285.06
also known as H53285-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120153-08-4
IUPAC Name
4-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Formula
C7H6BFO4
InChI Key
CZDWJVSOQOMYGC-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(C(O)=O)C(F)=C1
Assay (HPLC)
>97.5%

Description

4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text