Thermo Scientific Chemicals

4-Bromo-1-butyne, 97%, Thermo Scientific Chemicals

Catalog number: H53367.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-1-butyne, 97%, Thermo Scientific Chemicals

Catalog number: H53367.14
25 g, Each
Quantity
Catalog number: H53367.14
also known as H53367-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38771-21-0
IUPAC Name
4-bromobut-1-yne
Molecular Formula
C4H5Br
InChI Key
XLYOGWXIKVUXCL-UHFFFAOYSA-N
SMILES
BrCCC#C
Form
Liquid
Appearance (Color)
Colorless to pale brown
Assay (GC)
>97%

Description

4-Bromo-1-butyne is used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1-butyne is used for pharmaceutical intermediates.

Solubility
Immiscible with water.

Notes
Recommended storage in refrigerator. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text