Thermo Scientific Chemicals

2-Methyl-1-pentene, 97%, Thermo Scientific Chemicals

Catalog number: H53373.AC
25 mL, Each
Thermo Scientific Chemicals

2-Methyl-1-pentene, 97%, Thermo Scientific Chemicals

Catalog number: H53373.AC
25 mL, Each
Quantity
Catalog number: H53373.AC
also known as H53373-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
763-29-1
IUPAC Name
2-methylpent-1-ene
Molecular Formula
C6H12
InChI Key
WWUVJRULCWHUSA-UHFFFAOYSA-N
SMILES
CCCC(C)=C
Assay (HPLC)
96.0% min

Description

2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text