Thermo Scientific Chemicals

1-Iodo-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: H53375.06
5 g, Each
Thermo Scientific Chemicals

1-Iodo-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: H53375.06
5 g, Each
Quantity
Catalog number: H53375.06
also known as H53375-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
18163-47-8
Assay (HPLC)
96.0% min

Description

1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Vapors
    Category 4
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Reproductive Toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H361 - Suspected of damaging fertility or the unborn child

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention