Thermo Scientific Chemicals

1-(1-Adamantyl)ethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H53377.06
5 g, Each
Thermo Scientific Chemicals

1-(1-Adamantyl)ethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H53377.06
5 g, Each
Quantity
Catalog number: H53377.06
also known as H53377-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1501-84-4
Appearance (Color)
White
Assay from Supplier's CofA
≥97.5%
Form
Crystalline powder

Description

1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

Solubility
Freely soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text