N(alpha)-Boc-L-arginine, 98%, Thermo Scientific Chemicals
N(alpha)-Boc-L-arginine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N(alpha)-Boc-L-arginine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H53379.06
also known as H53379-06
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
Add to cart
N(alpha)-Boc-L-arginine, 98%, Thermo Scientific Chemicals
Catalog numberH53379.06
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13726-76-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[(diaminomethylidene)amino]pentanoic acid
Molecular FormulaC11H22N4O4
InChI KeyHSQIYOPBCOPMSS-ZETCQYMHSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis≥94% (on nitrogen content basis)
Elemental AnalysisC: 46.0 ? 1%, H: 8.0 ? 1%. N: 19.0 ? 1% Calc. for x 0.65 H₂O (4%)
Water Content (Karl Fischer Titration)≤5.0%
Optical Rotation|1²⁴ = -6 ? 2? (C=2 in AcOH)
Solution TestClear colorless solution (50 mg/mL in Water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only