Thermo Scientific Chemicals

3-(Trimethylsilyl)propiolic acid, 97%, Thermo Scientific Chemicals

Catalog number: H53380.06
5 g, Each
Thermo Scientific Chemicals

3-(Trimethylsilyl)propiolic acid, 97%, Thermo Scientific Chemicals

Catalog number: H53380.06
5 g, Each
Quantity
Catalog number: H53380.06
also known as H53380-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5683-31-8
Purity
≥96.0%
Appearance (Color)
White to pale yellow
Form
Solid or liquid
Infrared spectrum
Conforms

Description

3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text