2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Manuka Ghosal,; Sukanta Bhattacharyya,; Debabrata Mukherjee.Total synthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether. Tetrahedron Letters. 1989, 30(26),3469-3470.
- Andrew S. Kende,; Bruce Roth,; Pauline J. Sanfilippo. Facile, palladium(II)-mediated synthesis of bridged and spirocyclic bicycloalkenones. J. Am. Chem. Soc. 1982, 104 (6), 1784-1785.