Thermo Scientific Chemicals

1-Chloro-5-trimethylsilyl-4-pentyne, 97%, Thermo Scientific Chemicals

Catalog number: H53393.14
25 g, Each
Thermo Scientific Chemicals

1-Chloro-5-trimethylsilyl-4-pentyne, 97%, Thermo Scientific Chemicals

Catalog number: H53393.14
25 g, Each
Quantity
Catalog number: H53393.14
also known as H53393-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
77113-48-5
Assay from Suppliers CofA
≥96.0%
Appearance (Color)
Colorless to yellow
Form
Liquid

Description

1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    EU Specific Hazard Statements

    Combustible liquid