5-Decyn-1-ol, 97%, Thermo Scientific Chemicals
5-Decyn-1-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Decyn-1-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53400.06
also known as H53400-06
Price (USD)/ Each
79.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
79.60
Add to cart
5-Decyn-1-ol, 97%, Thermo Scientific Chemicals
Catalog numberH53400.06
Price (USD)/ Each
79.60
-
Add to cart
Specifications
Chemical Name or Material5-Decyn-1-ol
CAS68274-97-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Pei Nian Liu Dr.,Fu Hai Su Dr.,; Ting Bin Wen Dr.,; Herman H.-Y. Sung Dr.,; Ian D. Williams Prof. Dr., Guochen Jia Prof. Dr. Selective and Efficient Cycloisomerization of Alkynols Catalyzed by a New Ruthenium Complex with a Tetradentate Nitrogen-Phosphorus Mixed Ligand. TOC 2010, 16 (26) ,7889-7897.