2',3'-O-Isopropylideneadenosine, 98%, Thermo Scientific Chemicals
2',3'-O-Isopropylideneadenosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',3'-O-Isopropylideneadenosine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53401.06
also known as H53401-06
Price (USD)/ Each
61.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
61.40
Add to cart
2',3'-O-Isopropylideneadenosine, 98%, Thermo Scientific Chemicals
Catalog numberH53401.06
Price (USD)/ Each
61.40
-
Add to cart
Chemical Identifiers
CAS362-75-4
IUPAC Name[(3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]methanol
Molecular FormulaC13H17N5O4
InChI KeyLCCLUOXEZAHUNS-AUWRGFAENA-N
SMILESCC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
2',3'-O-Isopropylideneadenosine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,3′-O-Isopropylideneadenosine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Peter J.Harper; Alexander Hampton. Conversion of 2',3'-O-isopropylideneadenosine into the 5',5'-di-C-methyl derivative. J. Org. Chem. 1970, 35, (5),1688-1689
  2. Kentaro Anzai; Jun Uzawa. Cyclonucleoside formation and ring cleavage in the reaction of 2',3'-O-isopropylideneadenosine with benzoyl chloride and its substituted derivatives. J. Org. Chem. 1984, 49, (26),5076-5080