(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Shawn K. Collins,; Glenn P. A. Yap; Alex G. Fallis. A Novel Strained Undecadiyne Cyclophane with Interesting Dienophilic Character. ORGANIC LETTERS. 2001 , (1), 11-14.
Rolf W. Saalfran ; Martin Haubner,; Christian Deutscher,; Walter Bauer.; Timothy Clark.Cation-Mediated, Substituent-Controlled, C2-C7 Cycloaromatization of an Enyne-Allen. J. Org. Chem.. year of publication , 64 (17), 6166-6168.