Thermo Scientific Chemicals

N-Boc-propargylamine, 97%, Thermo Scientific Chemicals

Catalog number: H53406.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-propargylamine, 97%, Thermo Scientific Chemicals

Catalog number: H53406.06
5 g, Each
Quantity
Catalog number: H53406.06
also known as H53406-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92136-39-5
IUPAC Name
tert-butyl N-(prop-2-yn-1-yl)carbamate
Molecular Formula
C8H13NO2
InChI Key
DSPYCWLYGXGJNJ-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCC#C
Appearance (Color)
Yellow to brown
Form
Solid
Assay (GC)
>97%
Identification (FTIR)
Conforms to structure

Description

N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text