Thermo Scientific Chemicals

DL-Isoserine, 98%, Thermo Scientific Chemicals

Catalog number: H53412.03
1 g, Each
Thermo Scientific Chemicals

DL-Isoserine, 98%, Thermo Scientific Chemicals

Catalog number: H53412.03
1 g, Each
Quantity
Catalog number: H53412.03
also known as H53412-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
565-71-9
IUPAC Name
3-amino-2-hydroxypropanoic acid
Molecular Formula
C3H7NO3
InChI Key
BMYNFMYTOJXKLE-UHFFFAOYNA-N
SMILES
NCC(O)C(O)=O
Appearance (Color)
White
Assay (unspecified)
>97.5%
Elemental Analysis
<13.1% Nitrogen
Elemental Analysis
<33.71% Carbon
Elemental Analysis
<6.79% Hydrogen

Description

DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

Solubility
Soluble in HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text