DL-Isoserine, 98%, Thermo Scientific Chemicals
DL-Isoserine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Isoserine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H53412.06
also known as H53412-06
Price (USD)/ Each
695.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
695.00
Add to cart
DL-Isoserine, 98%, Thermo Scientific Chemicals
Catalog numberH53412.06
Price (USD)/ Each
695.00
-
Add to cart
Chemical Identifiers
CAS565-71-9
IUPAC Name3-amino-2-hydroxypropanoic acid
Molecular FormulaC3H7NO3
InChI KeyBMYNFMYTOJXKLE-UHFFFAOYNA-N
SMILESNCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)>97.5%
Elemental Analysis<13.1% Nitrogen
Elemental Analysis<33.71% Carbon
Elemental Analysis<6.79% Hydrogen
View more
DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

Solubility
Soluble in HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Theresa M. Williams,; Robyn Crumbie,; Harry S. Mosher. dl-Isoserine and related compoundsdl-Isoserine and related compounds . J. Org. Chem. 1985 , 50 (1), 91-97.
  2. Antonio Braibanti,; Francesco Dallavalle,; Giovanni Mori,; Luciano Valla.Thermodynamics of complex formation: DL-isoserine with nickel(II) and copper(II). Inorganica Chimica Acta. 1980, 40 70.