Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Alan R.Healy; Dr. Miho Izumikawa; Prof. Alexandra M.Z.Slawin; Dr. Kazuo Shin-ya; Prof. Nicholas J.Westwood. Stereochemical assignment of the protein-protein interaction inhibitor JBIR-22 by total synthesis . Angewandte Chemie International Edition. 2015, 54, (13),4046-4050