Thermo Scientific Chemicals

Cyclohexylacetylene, 98%, Thermo Scientific Chemicals

Catalog number: H53418.14
25 g, Each
Thermo Scientific Chemicals

Cyclohexylacetylene, 98%, Thermo Scientific Chemicals

Catalog number: H53418.14
25 g, Each
Quantity
Catalog number: H53418.14
also known as H53418-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
931-48-6
IUPAC Name
ethynylcyclohexane
Molecular Formula
C8H12
InChI Key
SSDZYLQUYMOSAK-UHFFFAOYSA-N
SMILES
C#CC1CCCCC1
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
>97.5%

Description

Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text