Specifications
Boiling Point135°C
Chemical Name or Material1-Trimethylsilyl-1-pentyne
CAS18270-17-2
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more