Thermo Scientific Chemicals

sec-Butyl methyl ether, 99%, Thermo Scientific Chemicals

Catalog number: H53427.AE
100 mL, Each
Thermo Scientific Chemicals

sec-Butyl methyl ether, 99%, Thermo Scientific Chemicals

Catalog number: H53427.AE
100 mL, Each
Quantity
Catalog number: H53427.AE
also known as H53427-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6795-87-5
IUPAC Name
2-methoxybutane
Molecular Formula
C5H12O
InChI Key
FVNIMHIOIXPIQT-UHFFFAOYNA-N
SMILES
CCC(C)OC
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥98.5%

Description

sec-Butyl methyl ether is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text