N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals
N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H53430.06
also known as H53430-06
Price (USD)/ Each
30.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
30.70
Add to cart
N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals
Catalog numberH53430.06
Price (USD)/ Each
30.70
-
Add to cart
Chemical Identifiers
CAS21678-37-5
IUPAC NameN,N,2-trimethylpropanamide
Molecular FormulaC6H13NO
InChI KeyGXMIHVHJTLPVKL-UHFFFAOYSA-N
SMILESCC(C)C(=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
1- chloro-N,N-2-trimethylpropenylamine readily prepared' from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jose Toye,; Leon Ghosez. Synthesis and reactivity of .alpha.-cyanoenamines. Novel general method for preparing .alpha.-diketones from amides J. Am. Chem. Soc.. 1975, 97 (8), 2276-2277.
  2. L. N. Domelsmith,; Linda L. Munchausen,; K. N. Houk.Lysergic acid diethylamide. Photoelectron ionization potentials as indices of behavioral activity. J. Med. Chem., 1977, 20 (10), 1346-1348.