Thermo Scientific Chemicals

N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals

Catalog number: H53430.22
100 g, Each
Thermo Scientific Chemicals

N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals

Catalog number: H53430.22
100 g, Each
Quantity
Catalog number: H53430.22
also known as H53430-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21678-37-5
IUPAC Name
N,N,2-trimethylpropanamide
Molecular Formula
C6H13NO
InChI Key
GXMIHVHJTLPVKL-UHFFFAOYSA-N
SMILES
CC(C)C(=O)N(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%

Description

1- chloro-N,N-2-trimethylpropenylamine readily prepared' from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text