Thermo Scientific Chemicals

N-(4-Pentynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: H53432.06
5 g, Each
Thermo Scientific Chemicals

N-(4-Pentynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: H53432.06
5 g, Each
Quantity
Catalog number: H53432.06
also known as H53432-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6097-07-0
IUPAC Name
2-(pent-4-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C13H11NO2
InChI Key
YNZIPXLLPFYDGM-UHFFFAOYSA-N
SMILES
O=C1N(CCCC#C)C(=O)C2=CC=CC=C12
Assay (GC)
≥96.0%
Appearance (Color)
White to yellow to pale brown
Form
Powder

Description

N-(4-Pentynyl)phthalimide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Pentynyl)phthalimide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text