1-Phenyl-1-hexyn-3-ol, 97%, Thermo Scientific Chemicals
1-Phenyl-1-hexyn-3-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenyl-1-hexyn-3-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53437.06
also known as H53437-06
Price (USD)/ Each
69.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
69.90
Add to cart
1-Phenyl-1-hexyn-3-ol, 97%, Thermo Scientific Chemicals
Catalog numberH53437.06
Price (USD)/ Each
69.90
-
Add to cart
Specifications
Chemical Name or Material1-Phenyl-1-hexyn-3-ol
CAS1817-51-2
Recommended StorageAmbient temperatures
Density0.949
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only