Cyclopropylacetylene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Marlin D.Harmony; R.N.Nandi; James V.Tietz; Jong-In Choe; Stephen J.Getty; Stuart W.Staley. Microwave structures of cyanocyclopropane and cyclopropylacetylene. Effects of cyclopropyl .pi. conjugation on structure. J. Am. Chem. Soc. 1983, 105, (12),3947-3951
Thomas G.Back; K. Raman Muralidharan. Free-radical selenosulfonation of vinylcyclopropanes, a cyclopropylidene, and cyclopropylacetylene. Relative rates of chain-transfer, ring-opening, and inversion in radical intermediates. J. Org. Chem. 1989, 54, (1),121-125