Propargyl benzoate, 98%, Thermo Scientific Chemicals
Propargyl benzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propargyl benzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H53444.14
also known as H53444-14
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
Propargyl benzoate, 98%, Thermo Scientific Chemicals
Catalog numberH53444.14
Price (USD)/ Each
165.00
-
Add to cart
Specifications
Chemical Name or MaterialPropargyl benzoate
CAS6750-04-5
Recommended StorageAmbient temperatures
Density1.106
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only