cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Boo-Gyo Shin; Myoung-Souk Jang; Do Yeung Yoon; Walter Heitz. Vinyl-type polymerization of norbornene dicarboxylic acid dialkyl esters . Macromolecular Rapid Communications. 2004, 25, (6),728-732
Lixia Ren; Jiuyang Zhang; Xiaolong Bai; Christopher G.Hardy; Ken D.Shimizu; Chuanbing Tang. Preparation of cationic cobaltocenium polymers and block copolymers by “living” ring-opening metathesis polymerization. Chem. Sci. 2012, 3, (2),580-583