Thermo Scientific Chemicals

11-Chloro-1-undecene, 97%, Thermo Scientific Chemicals

Catalog number: H53449.AC
25 mL, Each
Thermo Scientific Chemicals

11-Chloro-1-undecene, 97%, Thermo Scientific Chemicals

Catalog number: H53449.AC
25 mL, Each
Quantity
Catalog number: H53449.AC
also known as H53449-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-17-3
IUPAC Name
11-chloroundec-1-ene
Molecular Formula
C11H21Cl
InChI Key
PSEVKFKRYVAODC-UHFFFAOYSA-N
SMILES
ClCCCCCCCCCC=C
Form
Liquid
Assay from Supplier's CofA
≥96.0%
Appearance (Color)
Colorless to pale yellow
Infrared spectrum
Conforms

Description

11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text