4-Ethynylpyridine hydrochloride, 97%, Thermo Scientific Chemicals
4-Ethynylpyridine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethynylpyridine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53455.03
also known as H53455-03
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
4-Ethynylpyridine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH53455.03
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS352530-29-1
IUPAC Name4-ethynylpyridine hydrochloride
Molecular FormulaC7H6ClN
InChI KeyKFBZWZGIZHLUBX-UHFFFAOYSA-N
SMILESCl.C#CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormPowder
Assay (Freebase GC)>96.0%
4-pyridylacetylene hydrochloride is used in the preparation of alkynyl platinum(II) compounds with dichloro (1,2-bis(diphenylphosphino)ethane)platinium(II) complex in the presence of base and copper iodide as catalyst. It is used as an intermediate in the synthesis of 1,4-bis(4-pyridyl)butadiyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-pyridylacetylene hydrochloride is used in the preparation of alkynyl platinum(II) compounds with dichloro (1,2-bis(diphenylphosphino)ethane)platinium(II) complex in the presence of base and copper iodide as catalyst. It is used as an intermediate in the synthesis of 1,4-bis(4-pyridyl)butadiyne.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Mishra, A.; Ravikumar, S.; Song, Y. H.; Prabhu, N. S.; Kim, H.; Hong, S. H.; Cheon, S.; Noh, J.; Chi, K. W. A new arene-Ru based supramolecular coordination complex for efficient binding and selective sensing of green fluorescent protein. Dalton Trans. 2014, 43 (16), 6032-6040.
  2. Alvarino, C.; Simond, D.; Lorente, P. M.; Besnard, C.; Williams, A. F. Chains, Necklaces and Weaving Chain-link Grids from Self-Assembly Reactions. Chem. Eur. J. 2015, 21 (24), 8851-8858.