Tripropargylamine, 97%, Thermo Scientific Chemicals
Tripropargylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tripropargylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H53459.06
also known as H53459-06
Price (USD)/ Each
154.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
154.00
Add to cart
Tripropargylamine, 97%, Thermo Scientific Chemicals
Catalog numberH53459.06
Price (USD)/ Each
154.00
-
Add to cart
Specifications
Chemical Name or MaterialTripropargylamine
CAS6921-29-5
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
Tripropargylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripropargylamine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sachin A.Ingale; Frank Seela. A ratiometric fluorescent on-off Zn2+ chemosensor based on a tripropargylamine pyrene azide click adduct. J. Org. Chem. 2012, 77, (20),9352-9356
  2. Yeong-Soon Gal; Won-Chul Lee; Sam-Kwon Choi; Yun-Chul Kim; Bal Jung. Polymerization of tripropargylamine by transition metal catalysts and molecular structure of poly(tripropargylamine). European Polymer Journal. 1996, 32, (5),579-585