Specifications
Chemical Name or Material1,1,3-Triphenylpropargyl alcohol
CAS1522-13-0
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more