3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Ethynylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53465.03
also known as H53465-03
Price (USD)/ Each
65.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.30
Add to cart
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
Catalog numberH53465.03
Price (USD)/ Each
65.30
-
Add to cart
Chemical Identifiers
CAS2510-23-8
IUPAC Name3-ethynylpyridine
Molecular FormulaC7H5N
InChI KeyCLRPXACRDTXENY-UHFFFAOYSA-N
SMILESC#CC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to light brown
FormSolid
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only