4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals
4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H53466.14
also known as H53466-14
Price (USD)/ Each
386.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
386.00
Add to cart
4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals
Catalog numberH53466.14
Price (USD)/ Each
386.00
-
Add to cart
Chemical Identifiers
CAS936-58-3
IUPAC Name1-phenylbut-3-en-1-ol
Molecular FormulaC10H12O
InChI KeyRGKVZBXSJFAZRE-UHFFFAOYNA-N
SMILESOC(CC=C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay (GC)>96%
Identification (FTIR)Conforms to structure
4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Liang Xu; Mark L.Trudell. A mild and efficient oxidation of alcohols to aldehydes and ketones with periodic acid catalyzed by chromium(III) acetylacetonate. Tetrahedron Letters. 2003, 44, (12),2553-2555
  2. John Fawcett; Eric G.Hope; Alison M.Stuart; Andrew J.West. Recycling of a perfluoroalkylated BINOL ligand using fluorous solid-phase extraction. Green Chem. 2005, 7, (5),316-320