Thermo Scientific Chemicals

4,6-Decadiyne, 97%, Thermo Scientific Chemicals

Catalog number: H53467.03
1 g, Each
Thermo Scientific Chemicals

4,6-Decadiyne, 97%, Thermo Scientific Chemicals

Catalog number: H53467.03
1 g, Each
Quantity
Catalog number: H53467.03
also known as H53467-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16387-71-6
IUPAC Name
deca-4,6-diyne
Molecular Formula
C10H14
InChI Key
LIWZSNTUMSGWTF-UHFFFAOYSA-N
SMILES
CCCC#CC#CCCC
Appearance (Color)
Colorless to yellow to brown
Form
Liquid
Assay from Supplier's CofA
≥96.0%

Description

6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

Solubility
Immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text