Thermo Scientific Chemicals

Propargyl ether, 98%, Thermo Scientific Chemicals

Catalog number: H53473.14
25 g, Each
Thermo Scientific Chemicals

Propargyl ether, 98%, Thermo Scientific Chemicals

Catalog number: H53473.14
25 g, Each
Quantity
Catalog number: H53473.14
also known as H53473-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6921-27-3
IUPAC Name
3-(prop-2-yn-1-yloxy)prop-1-yne
Molecular Formula
C6H6O
InChI Key
HRDCVMSNCBAMAM-UHFFFAOYSA-N
SMILES
C#CCOCC#C
Form
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Colorless to yellow
Assay from Supplier's CofA
≥97.5%

Description

Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1?-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1′-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text