Acetophenone p-toluenesulfonylhydrazone, 98%, Thermo Scientific Chemicals
Acetophenone p-toluenesulfonylhydrazone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetophenone p-toluenesulfonylhydrazone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53474.06
also known as H53474-06
Price (USD)/ Each
64.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.90
Add to cart
Acetophenone p-toluenesulfonylhydrazone, 98%, Thermo Scientific Chemicals
Catalog numberH53474.06
Price (USD)/ Each
64.90
-
Add to cart
Specifications
Chemical Name or MaterialAcetophenone p-toluenesulfonylhydrazone
CAS4545-21-5
Melting Point144°C to 149°C
Recommended StorageAmbient temperatures
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only