Thermo Scientific Chemicals

Cyclopentylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: H53483.06
5 g, Each
Thermo Scientific Chemicals

Cyclopentylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: H53483.06
5 g, Each
Quantity
Catalog number: H53483.06
also known as H53483-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
930-51-8
IUPAC Name
ethynylcyclopentane
Molecular Formula
C7H10
InChI Key
TXVJSWLZYQMWPC-UHFFFAOYSA-N
SMILES
C#CC1CCCC1
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay (GC)
≥ 96.0%

Description

Cyclopentylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetylene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text