3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals
3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H53484.06
also known as H53484-06
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
Add to cart
3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals
Catalog numberH53484.06
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1671-87-0
IUPAC Namebis(pyridin-2-yl)-1,2,4,5-tetrazine
Molecular FormulaC12H8N6
InChI KeyJFBIRMIEJBPDTQ-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to dark red to purple
FormCrystalline powder
Assay from Supplier's CofA≥95%
3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Xian-He Bu; Hiromasa Morishita; Kentaro Tanaka; Kumar Biradha; Sanae Furusho; Mitsuhiko Shionoya. A spontaneously resolved chiral molecular box: a cyclic tetranuclear ZnIIcomplex with DPTZ (DPTZ = 3,6-di-2-pyridyl-1,2,4,5-tetrazine) . Chem. Commun. 2000, (11),971-972
  2. Jiehu Cui; Liangfang Huang; Zhenzhong Lu; Yizhi Li; Zijian Guo; Hegen Zheng. Synthesis and properties of five unexpected copper complexes with ring-cleavage of 3,6-di-2-pyridyl-1,2,4,5-tetrazine by one pot in situ hydrothermal reaction. CrystEngComm. 2012, 14, (6),2258-2267