Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H53487.14
also known as H53487-14
Price (USD)/ Each
404.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
404.00
Add to cart
Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Catalog numberH53487.14
Price (USD)/ Each
404.00
-
Add to cart
Specifications
Boiling Point83°C (80mmHg)
Chemical Name or MaterialCyclopropyl(trimethylsilyl)acetylene
CAS81166-84-9
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

Solubility
Sparingly Soluble (0.069 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only