Thermo Scientific Chemicals

N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: H53489.06
5 g, Each
Thermo Scientific Chemicals

N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: H53489.06
5 g, Each
Quantity
Catalog number: H53489.06
also known as H53489-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6097-08-1
IUPAC Name
2-(hex-5-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C14H13NO2
InChI Key
WALVSJIWVWDFCU-UHFFFAOYSA-N
SMILES
O=C1N(CCCCC#C)C(=O)C2=CC=CC=C12
Assay (unspecified)
≥96.0%

Description

6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text