Specifications
Chemical Name or Material5-Methyl-1-hexyn-3-ol
CAS61996-79-0
Health Hazard 1H226-H302
Health Hazard 2GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
View more