Thermo Scientific Chemicals

Propargylamine, 98%, Thermo Scientific Chemicals

Catalog number: H53495.06
5 g, Each
Thermo Scientific Chemicals

Propargylamine, 98%, Thermo Scientific Chemicals

Catalog number: H53495.06
5 g, Each
Quantity
Catalog number: H53495.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2450-71-7
IUPAC Name
prop-2-yn-1-aminium chloride
Molecular Formula
C3H6ClN
InChI Key
IKXNIQJDNKPPCH-UHFFFAOYSA-N
SMILES
[Cl-].[NH3+]CC#C
Assay (GC)
>97.5

Description

Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Hygroscopic. Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text