Thermo Scientific Chemicals

Rubrene, 97%, Thermo Scientific Chemicals

Catalog number: H53507.14
25 g, Each
Thermo Scientific Chemicals

Rubrene, 97%, Thermo Scientific Chemicals

Catalog number: H53507.14
25 g, Each
Quantity
Catalog number: H53507.14
also known as H53507-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
517-51-1
IUPAC Name
5,6,11,12-tetraphenyltetracene
Molecular Formula
C42H28
InChI Key
YYMBJDOZVAITBP-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=C2C(C3=CC=CC=C3)=C3C=CC=CC3=C(C3=CC=CC=C3)C2=C(C2=CC=CC=C2)C2=CC=CC=C12
Appearance (Color)
Red
Form
Crystalline solid
Assay (HPLC)
>97%

Description

Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

Solubility
Soluble in hot toluene. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text