3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Yanchi Tang; Tiebo Xiao; Lei Zhou. Cobalt-catalyzed alkyne-dihalomethane-amine coupling: an efficient route for propargylamines. Tetrahedron Letters. 2012, 53, (46),6199-6201
Herman G.RicheyJr.; L.Meredith Moses; M.S.Domalski; Wayne F.Erickson; Alan S.Heyn. Reactions of allylic Grignard reagents and unsaturated amines. J. Org. Chem. 1981, 46, (19),3773-3780