1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Cristiana Cravo-Laureau; Agnès Hirschler-Réa; Robert Matheron; Vincent Grossi. Growth and cellular fatty-acid composition of a sulfate-reducing bacterium, Desulfatibacillum aliphaticivorans strain CV2803T, grown on n-alkenes. Comptes Rendus Biologies. 2004, 327 (7), 687-694.
R.A. Wirtz; S.L. Taylor; H.G. Semey. Concentrations of substituted p-benzoquinones and 1-pentadecene in the flour beetles tribolium confusum J. Du Val and tribolium castaneum (herbst). Comparative Biochemistry and Physiology Part B: Comparative Biochemistry. 1978, 61 (1), 25-28.