6-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
6-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromo-1-indanone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H54003.06
also known as H54003-06
Price (USD)/ Each
206.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
206.00
Add to cart
6-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
Catalog numberH54003.06
Price (USD)/ Each
206.00
-
Add to cart
Specifications
Chemical Name or Material6-Bromo-1-indanone
CAS14548-39-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point110°C
View more
6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Julius J.Matasi; John P.Caldwell; Jinsong Hao; Bernard Neustadt; Leyla Arik; Carolyn J.Foster; Jean Lachowicz; Deen B.Tulshian. The discovery and synthesis of novel adenosine receptor (A2A) antagonists. Bioorganic & Medicinal Chemistry Letters. 2005, 15, (5),1333-1336