3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
N. Subhakara Reddy; A.Srinivas Rao; M.Adharvana Chari; V.Ravi kumar; V.Jyothy; V.Himabindu. Synthesis and antibacterial activity of sulfonamide derivatives of anacardicacid mixture isolated from a natural product cashew nut shell liquid (CNSL). Letters in Organic Chemistry. 2012, 9, (4),287-293