Thermo Scientific Chemicals
Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog number: H54053.03
1 g, Each
Thermo Scientific Chemicals
Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog number: H54053.03
1 g, Each
Quantity
Catalog number: H54053.03
also known as H54053-03
Price (USD)
Quantity
-
Have Questions?
Description
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.