Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H54053.06
also known as H54053-06
Price (USD)/ Each
535.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
535.00
Add to cart
Piperidine-1-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH54053.06
Price (USD)/ Each
535.00
-
Add to cart
Specifications
Chemical Name or MaterialPiperidine-1-sulfonyl chloride
CAS35856-62-3
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shuyou Chen; Kui Zheng; Fan Chen. Palladium-copper-catalyzed desulfitative amination of benzo[d]oxazole C-H bond. Tetrahedron Letters. 2012, 53, (46),6297-6299
  2. Maurizio Tamba; Katalin Dajka; Carla Ferreri; Klaus-Dieter Asmus; Chryssostomos Chatgilialoglu. One-electron reduction of methanesulfonyl chloride. The fate of MeSO2Cl·- and MeSO2· intermediates in oxygenated solutions and their role in the cis-trans isomerization of mono-unsaturated fatty acids. J. Am. Chem. Soc. 2007, 129, (28),8716-8723