3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number H54057.AE
also known as H54057-AE
Price (USD)/ Each
522.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
522.00
Add to cart
3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
Catalog numberH54057.AE
Price (USD)/ Each
522.00
-
Add to cart
Specifications
Chemical Name or Material3,5-Dimethylphenylmagnesium bromide
CAS34696-73-6
Health Hazard 1H225-H302-H314-H335
Health Hazard 2GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
3,5-Dimethylphenylmagnesium bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylmagnesium bromide is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Hidehiko Sasaki; Ryo Irie; Tetsuya Hamada; Kenji Suzuki; Tsutomu Katsuki. Rational design of Mn-salen catalyst (2): Highly enantioselective epoxidation of conjugated cis olefins. Tetrahedron. 1994, 50, (41),11827-11838
  2. Derick D.Winkle; Kevin M.Schaab. Suzuki reaction of a diarylborinic acid: One-pot preparation and cross-coupling of bis(3,5-dimethylphenyl)borinic acid. Org. Proc. Res. Dev. 2001, 5, (4),450-451