4'-(Methylsulfonyl)acetophenone, 97+%, Thermo Scientific Chemicals
4'-(Methylsulfonyl)acetophenone, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-(Methylsulfonyl)acetophenone, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H54072.06
also known as H54072-06
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
4'-(Methylsulfonyl)acetophenone, 97+%, Thermo Scientific Chemicals
Catalog numberH54072.06
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS10297-73-1
IUPAC Name1-(4-methanesulfonylphenyl)ethan-1-one
Molecular FormulaC9H10O3S
InChI KeyKAVZYDHKJNABPC-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)S(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only